Lithium triethylborohydride-Lösung 1M in THF

Lieferant: Thermo Fisher Scientific

176991000 176998000
176991000.EA 91.3 EUR
176991000. 176998000.
Lithium triethylborohydride-Lösung 1M in THF
Lithium triethylborohydride
Formel: C₆H₁₆BLi
Dichte: 9200 g/cm³
MDL: MFCD00011703
CAS-Nummer: 22560-16-3
UN: 3148
ADR: 4.3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR