Supelco® Organic Reference Standard, Triglyceride Mix
Lieferant: Merck
17810-1AMP-S
17810-1AMP-SEA
126
EUR
17810-1AMP-S
Supelco® Organic Reference Standard, Triglyceride Mix
Standards
Organische Standards
Verpackung: 100 mg ampule contains approximately equal amounts by weight of: Triacetin (C2:0) Tributyrin (C4:0) Tricaproin (C6:0) Tricaprylin (C8:0) Tricaprin (C10:0)