4'-Hydroxy-3'-methoxyacetophenon, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Apocynin, Acetovanillon
178385-1
178385-1EA
114
EUR
178385-1
4'-Hydroxy-3'-methoxyacetophenon, Millipore®
4'-Hydroxy-3'-methoxyacetophenon
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Siedepunkt: 295 °C (1013 hPa) Schmelzpunkt: 115 °C |
MDL:
MFCD00008747 CAS-Nummer: 498-02-2 EINECS: 207-854-5 Merck Index: 13,00746 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...