Ammoniumformiat 99,0 Anhydrous basis HPLC, Fluka™
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Ameisensäure Ammoniumsalz
17843-250G
17843-50G
17843-250GEA
402.77
EUR
17843-250G
17843-50G
Ammoniumformiat 99,0 Anhydrous basis HPLC, Fluka™
Ammoniumformiat
Formel:
HCO₂NH₄ Molecular Weight: 63.06 g/mol Schmelzpunkt: 116 °C Dichte: 1.266 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013103 CAS-Nummer: 540-69-2 EINECS: 208-753-9 Merck Index: 13,00525 |
Spezifikation Testergebnisse
Appearance of the solution (c=10%) | complying |
Assay (calculated on the anhydrous basis) | 99.0 - 101.0 % |
Water (Karl Fischer) | Max. 2 % |
pH (10 %, 20°C) | 5.5 - 7.5 |
Absorbance at 250 nm | Max. 0.05 |
Absorbance at 260 nm | Max. 0.04 |
Absorbance at 270 nm | Max. 0.03 |
Absorbance 280 nm | Max. 0.02 |
Absorbance 500 nm | Max. 0.02 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...