CD437 ≥98% (durch HPLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: AHPN, 6-(3-(Adamantan-1-yl)-4-hydroxyphenyl)-2-naphthoic acid, Apoptosis Activator VI, 6-[3-(1-Adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

178496-5
178496-5EA 114 EUR
178496-5
CD437 ≥98% (durch HPLC), Calbiochem®, Sigma-Aldrich®
CD437

The Apoptosis Activator VI, CD437/AHPN, also referenced under CAS 125316-60-1, modulates Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.


A cell-permeable retinoid-related molecule that induces tumor growth arrest and apoptosis. The exact molecular basis of the underlying mechanism(s) is not yet well understood, however, studies using ovarian adenocarcinoma cells have shown that CD437/AHPN effectively functions as an ER stress inducer.

Formel: C₂₇H₂₆O₃
Molecular Weight: 398,4936 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03106506
CAS-Nummer: 125316-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR