Methylorange ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Goldorange, Helianthin, Natrium-4-(4-dimethylaminophenylazo)benzolsulfonat, Orange III, 4-(4-(Dimethylamino)phenylazo)benzolsulfonsäure Natriumsalz, (4-(Dimethylamino)azobenzol-4'-sulfonsäure Natriumsalz
17874.06
17874.14
17874.22
17874.06EA
22.1
EUR
17874.06
17874.14
17874.22
Methylorange ACS
Methylorange
CAS No.: 547-58-0
Molecular Formula: C14H14N3NaO3S
Formula Weight: 327.34
Physical Form: Crystalline
Appearance: Orange
Odour: Weak, characteristic
pH: 6.5
Melting Point:
Formel:
C₁₄H₁₄N₃NaO₃S Siedepunkt: ∼90 °C (1013 hPa) Schmelzpunkt: >300 °C Dichte: ∼1 g/cm³ (20 °C) Flammpunkt: 38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007502 CAS-Nummer: 547-58-0 EINECS: 208-925-3 UN: 3143 ADR: 6.1,III Merck Index: 14,6105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...