Aceton ≥99,5% ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

179124-6X1L 179124-1L 179124-2.5L 179124-4X2.5L
179124-6X1LEA 268.75 EUR
179124-6X1L 179124-1L 179124-2.5L 179124-4X2.5L
Aceton ≥99,5% ACS, Riedel-de Haën™
Aceton

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635680
RTECS Number: AL3150000
UNSPSC Code: 12191602

Formel: CH₃COCH₃
Siedepunkt: 55…57 °C (1,013 hPa)
Schmelzpunkt: –95 °C
Dichte: 0,79 g/cm³ (20 °C)
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay according to ACS (GC) Min. 99.5 %
Solubility in water complying
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.5 %
Titrable Acid Max. 0.0003 meq/g
Titrable Base Max. 0.0006 meq/g
KMnO₄ reducing substances complying
Aldehydes (as HCHO) Max. 0.002 %
Methanol (GC) Max. 0.05 %
2-propanol (GC) Max. 0.05 %
APHA Max. 10
Vapor Pressure 184 mmHg ( 20 °C)
Vapor Density 2 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR