Cyclohexan ≥99% ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

179191-1L 179191-6X1L
179191-1LEA 82.04 EUR
179191-1L 179191-6X1L
Cyclohexan ≥99% ACS, Riedel-de Haën™
Cyclohexan

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504

Formel: C₆H₁₂
Siedepunkt: 81 °C (1,013 hPa)
Schmelzpunkt: 6,4 °C
Dichte: 0,78 g/cm³ (20 °C)
Flammpunkt: -18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Vapor Pressure 77 mmHg ( 20 °C)
IR spectrum complying
Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.02 %
Reaction against H₂SO₄ complying
UV-absorption complying
APHA Max. 10
Refractive Index n20/D 1.426 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR