Methanol 99,8 ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol

179337-1L 179337-500ML 179337-6X1L 179337-6X500ML
179337-1LEA 68.96 EUR
179337-1L 179337-500ML 179337-6X1L 179337-6X500ML
Methanol 99,8 ACS, Riedel-de Haën™
Methanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1098229
RTECS Number: PC1400000
UNSPSC Code: 12191601

Formel: CH₃OH
Molecular Weight: 32.04 g/mol
Siedepunkt: 64.7 °C
Schmelzpunkt: -98
Dichte: 0.792 g/cm³ (20 °C)
Flammpunkt: 11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay (GC) Min. 99.80 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.0003 meq/g
Titrable Base Max. 0.0002 meq/g
KMnO₄ reducing matter complying
Acetone (GC) Max. 0.001 %
Formaldehyde Max. 0.001 %
Acetaldehyde Max. 0.001 %
Mixable with H₂O complying
Reaction against H₂SO₄ complying
APHA Max. 10
Refractive Index n20/D 1.329 (lit.)
Vapor Density 1.11 (vs air)
Vapor Pressure 97.68 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR