Ninhydrin Reagens acc.to Stahl, gelbe Flüssigkeit Derivatisierungsreagenz, für DC
Lieferant: Merck
|
Gefahr
|
Synonyme:
2,2-Dihydroxy-1H-inden-1,3(2H)-dion, 2,2-Dihydroxy-1,3-indendion, Indantrionhydrat, 2,2-Dihydroxy-1,3-indandion, 2,2-Dihydroxyhydrinden-1,3-dion
M-Clarity™ Qualitätsstufe = MQ100
Ninhydrin reagent is mostly used for the detection of amino acids. It basically gives a characteristic purple color on reaction with a-amino acids and other amino compounds, provided the pH of the reaction is 5.0 or higher.
At an acid pH of 0.3, the reaction of ninhydrin with ornithinine and proline results in a red color product. Ninhydrin reagent can stay stable at room temperature in the absence of nitrogen atmosphere for upto 1 month in the ethylene glycol-sodium acetate solvent system.
Ninhydrin reagent may be used for the quantitative determination of total protein content in tissue hydrolysates.
Formel:
C₉H₆O₄ Molecular Weight: 178,14 g/mol Dichte: 0,823 g/ml (20 °C) |
MDL:
MFCD00003791 CAS-Nummer: 485-47-2 UN: 1120 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...