Triammoniumdodecamolybdophosphat Hydrat, Reagenzqualität
Lieferant: Thermo Fisher Scientific
|
Achtung
|
17984.22
17984.36
17984.22EA
75.4
EUR
17984.22
17984.36
Triammoniumdodecamolybdophosphat Hydrat, Reagenzqualität
Triammoniumdodecamolybdophosphat Hydrat
CAS No.: 54723-94-3
Molecular Formula: H12Mo12N3O40P.xH2O (x≈3)
Formula Weight: 1876.35(anhy)
Physical Form: Powder
Density: 3.15
Appearance: Yellow
Melting Point: dec.
MDL No.: MFCD00036312
Formel:
H₁₂Mo₁₂N₃O₄₀P·H₂O Dichte: 3,15 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150423 CAS-Nummer: 54723-94-3 EINECS: 234-714-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...