Ammoniumsulfit Monohydrat ≥92%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 92% min
CAS No.: 7783-11-1
Molecular Formula: H8N2O3S.H2O
Formula Weight: 134.15 (116.14anhy)
Physical Form: Crystalline
Density: 1.41
Refractive Index: 1.5150
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 60-70° dec.
Merck Reference: 14,557
MDL No.: MFCD00066094
Formel:
H₈N₂O₃S·1H₂O Dichte: 1,41 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066094 CAS-Nummer: 7783-11-1 EINECS: 233-484-9 Merck Index: 14,557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...