Chart recorder pens
Lieferant: Cytiva
18-1004-23
18-1004-23EA 217 EUR
18-1004-23
Chart recorder pens
Schreibstifte Stifte für Linienschreiber
Pens for use with paper chart recorders.
  • Response time of 0,2 s (2,55 to 97,5% FSD)
  • Fibre tip
Available in blue or red.