1-Naphthylbutyrat-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: α-Naphthylbutyrat, Buttersäure(1-naphthyl)ester, Buttersäure(α-naphthyl)ester

1801-50ML
1801-50MLEA 177 EUR
1801-50ML
1-Naphthylbutyrat-Lösung, Sigma-Aldrich®
1-Naphthylbutyrat

Intended for use in the Sigma α-Butyrate Esterase kit, procedure 181B

Formel: C₁₄H₁₄O₂
Molecular Weight: 214,26 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00003923
CAS-Nummer: 3121-70-8
ADR: 3,III
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR