Sulphur standard solutions for ICP, TraceCERT®, Supelco®
Lieferant: Merck
18021-100ML
18021-100MLEA
109
EUR
18021-100ML
Sulphur standard solutions for ICP, TraceCERT®, Supelco®
Standards
Kationischer Standards
Schwefel-Standards
These standard solutions are used for ICP and AAS.
Standards: Accordance with ISO/IEC 17025 and ISO 17034.