Pararosaniline-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base

1804-15ML
1804-15MLEA 194 EUR
1804-15ML
Pararosaniline-Lösung, Sigma-Aldrich®
Pararosaniline

When mixed with a 4% sodium nitrite solution, a freshly hexazotized pararosaniline solution is produced. May be used in a variety of cytochemistry procedures, including Sigma Procedures 181B and 181C.

Formel: C₁₉H₁₉N₃O
Molecular Weight: 305.38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036222
CAS-Nummer: 467-62-9
ADR: 8 II
IVDLogo

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR