Methylenblau Lösung R in Ethanol 95%

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

1808-50ML
1808-50MLEA 104 EUR
1808-50ML
Methylenblau Lösung R in Ethanol 95%
Methylenblau

Used for in vitro diagnostic. It is used as a nuclear counterstain in the various Lymphocyte enzyme kits, Procedure No. 181.

Formel: C₁₆H₁₈ClN₃S
Flammpunkt: 13 °C
MDL: MFCD00150008
CAS-Nummer: 61-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR