VWR® PharMed® BPT, Pump Tubing, Coiled
Lieferant: VWR Collection
181-0557EA 0 EUR
181-0557
VWR® PharMed® BPT, Pump Tubing, Coiled
Schläuche Transferschlauch
Catalyst pump coiled tubing pumps.
  • Four formulations available to cover most chemical compatible applications
  • Available in both thin and thick wall configurations
To help simplify choices, a platinum-cured silicone, two TPEs and a long life PVC tubing material are offered.