Absaugsysteme, Biovac 106
Lieferant: Welch by Gardner Denver
112037 112037-02 112037-04
181-0176EA 1950 EUR
181-0176 181-0587 181-0588
Absaugsysteme, Biovac 106
Pumpen Fass/Transferpumpen
With the biovac 106, nonflammable fluids can be removed easily and very precisely from slides, Petri dishes, cell culture containers, etc, by using different pipettes or glass tips and Pasteur pipettes which can be easily connected to the handvac pipette. The biovac 106 is switched on and off directly at the device or via the optionally available foot switch.
  • Resistant diaphragm pump
  • Operation via ON/OFF switch or optional foot switch
  • Autoclavable receiver is available in 2 L glass or 4 L polypropylene
  • Power supply: 115/230 V, 50/60 Hz
A float valve stops the bottle from being overfilled and protects the pump. A hydrophobic filter with a suitable filtration grade can be inserted for additional protection.
An ultra quiet (45 dBA), chemically resistant diaphragm pump can be activated by either the hand piece or optional foot switch. All included tubing is chemically resistant.
Power supply: 115V/230V 50/60Hz
Bestellinformation: Filters and accessories are sold separately. A wide range of available accessories and filters allow the user to customise the biovac 106 depending on their specific needs. Contact your local Avantor representative for more information.