Vacuum gauge
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
E-1120
181-9915EA 94.7 EUR
181-9915
Vacuum gauge
Pumpen Vakuum-Messgeräte
Simple brass vacuum/pressure gauge for inclusion in vacuum lines to give coarse vacuum/pressure indication.
  • Display: 1 to 0 bar (Vacuum) and 0 to 1,5 bar (Pressure)
  • Display Ø: 60 mm