Zinc citrate dihydrate (≥31% Zn), Reagenzqualität

Lieferant: Thermo Fisher Scientific

18129.A1
18129.A1EA 136 EUR
18129.A1
Zinc citrate dihydrate (≥31% Zn), Reagenzqualität
Zinc citrate dihydrate

CAS No.: 5990-32-9 Molecular Formula: C12H10O14Zn3.2H2O Formula Weight: 610.35 (574.32anhy) Physical Form: Powder Odour: Odourless Melting Point: 334° dec. Solubility: Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water MDL No.: MFCD00150587

Formel: C₁₂H₁₄O₁₆Zn₃
Molecular Weight: 610,368 g/mol
Schmelzpunkt: 334 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150587
CAS-Nummer: 5990-32-9
EINECS: 208-901-2
Merck Index: 14,10134

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR