Bismuth indium lead tin eutectic ingot (alloy 136) ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
18143.22
18143.22EA
166
EUR
18143.22
Bismuth indium lead tin eutectic ingot (alloy 136) ≥99,9% (Metall-Basis)
Bismuth indium lead tin eutectic ingot (alloy 136)
Purity: 99.9%
Molecular Formula: Bi:In:Pb:Sn; 49:21:18:12 wt%
Odour: Odourless
Melting Point: 58°
MDL No.: MFCD00210606
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...