α-Naphthyl Butyrate Esterase, Lymphocyte kit, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ500

181B-1KT
181B-1KTEA 344 EUR
181B-1KT
α-Naphthyl Butyrate Esterase, Lymphocyte kit, Sigma-Aldrich®
α-Naphthyl Butyrate Esterase, Lymphocyte kit

This kit is intended for demonstration of nonspecific esterase in a subset of lymphocytes for determining maturation arrest. Kits 90A1 or 91A are intended for monocyte identification.


  • Suitable for haematology
  • Suitable for histology


Demonstration of lymphocyte staining patterns may indicate specific stages of maturation arrest associated with certain lymphoproliferative disorders.


Kit component only
α-Naphthyl Butyrate Solution (180-1) 50 mL
Citrate Solution (91-5) 50 mL
Methylene Blue Solution (180-8) 50 mL
Pararosaniline Solution (180-4) 15 mL
Phosphate Buffer (180-5) 1 mL/vial
Sodium Nitrite (kit only) 10 tablets

Lagertemperatur: Kühlschrank
UN: 3316
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR