Ethylmagnesiumbromid ∼3.2 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®

Lieferant: Thermo Fisher Scientific
Gefahr

183531000 183538000
183531000.EA 97.7 EUR
183531000. 183538000.
Ethylmagnesiumbromid ∼3.2 M in 2-MeTHF (2-Methyltetrahydrofuran), AcroSeal®
Ethylmagnesium bromide
Formel: C₂H₅BrMg
Molecular Weight: 133.27 g/mol
Dichte: 1.14 g/cm³
Flammpunkt: –11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I

Bestellen

Spezifikation Testergebnisse

Appearance Dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 39 to 41 %
Water ≤1 %
Solubility (2 % in DMSO) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR