Spare seal rings for small flange connections
Lieferant: VACUUBRAND
20660110 20660111 20660115 20660116 20660130 20660135 20660136 20660137
185-1331EA 3.8 EUR
185-1331 185-1333 185-1332 185-1334 185-1341 185-1342 185-1344 185-1346
Spare seal rings for small flange connections
Versiegeln Dichtringe
These spare seal rings for small flange KF connections are made of NBR or FKM.