1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-EBIO, 1-Ethyl-1H-benzo[d]imidazol-2(3H)-one, 1-Ethylbenzimidazolinone

186310010
186310010.EA 101 EUR
186310010.
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one
Formel: C₉H₁₀N₂O
Molecular Weight: 162,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005715
CAS-Nummer: 10045-45-1
EINECS: 233-148-1

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR