Yttrium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
18682.18
18682.09
18682.30
18682.18EA
511
EUR
18682.18
18682.09
18682.30
Yttrium(III)chlorid, wasserfrei ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Yttrium(III)chlorid
Purity: 99.9%
CAS No.: 10361-92-9
Molecular Formula: Cl3Y
Formula Weight: 195.26
Physical Form: -20 Mesh
Density: 2.67
Appearance: White to pale yellow
Odour: Odourless
Melting Point: 721°
Merck Reference: 14,10107
MDL No.: MFCD00011470
Formel:
Cl₃Y Siedepunkt: 1488 °C (1013 hPa) Schmelzpunkt: 721 °C Dichte: 2,67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011470 CAS-Nummer: 10361-92-9 EINECS: 233-801-0 Merck Index: 14,10107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...