Zinc diethyldithiocarbamate (17 - 19,5% Zn)

Lieferant: Thermo Scientific
Achtung

Synonyme: Zinc dethyldithiocarbamate, Diethyldithiocarbamic Acid Zinc Salt

18697.36
18697.36EA 44.6 EUR
18697.36
Zinc diethyldithiocarbamate (17 - 19,5% Zn)
Zinc diethyldithiocarbamate

CAS No.: 14324-55-1 Molecular Formula: C10H20N2S4Zn Formula Weight: 361.90 Flash Point: 204°(400°F) Physical Form: Crystalline Density: 1.48 Appearance: Off-white Odour: Odourless pH: 7.8 Melting Point: 178-183° UN No.: UN3077 MDL No.: MFCD00064798


Applications: Chelating agent

Formel: C₅H₁₁NS₂
Molecular Weight: 361,90 g/mol
Schmelzpunkt: 178…183 °C
Dichte: 1,48
Flammpunkt: 204 °C (400 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064798
CAS-Nummer: 14324-55-1
EINECS: 238-270-9
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR