Bromwasserstoff ≥48%, Purum p.a.
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Bromwasserstoff, Hydrogenbromid
18730-1L
18730-1LEA
105.86
EUR
18730-1L
Bromwasserstoff ≥48%, Purum p.a.
Bromwasserstoff
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Formel:
HBr Molecular Weight: 80,91 g/mol Siedepunkt: 126 °C (1,013 hPa) Schmelzpunkt: -11 °C Dichte: 1,490 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011323 CAS-Nummer: 10035-10-6 EINECS: 233-113-0 UN: 1788 ADR: 8,II |
Spezifikation Testergebnisse
Assay | 48.0 - 50.0 % |
Refractive index (n 20/D) | 1.440 - 1.443 |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 5 ppm |
Zinc (Zn) | Max. 5 ppm |
Sulfate and sulfite (as SO₄) | Max. 50 ppm |
Vapor Pressure | 320 psi ( 21.1 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...