Iron(III) meso-tetraphenylporphine chloride
Lieferant: Thermo Fisher Scientific
18829.03
18829.MD
18829.03EA
214
EUR
18829.03
18829.MD
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride
CAS No.: 16456-81-8
Molecular Formula: C44H28ClFeN4
Formula Weight: 704.04
Physical Form: Crystalline
MDL No.: MFCD00012152
Formel:
C₄₄H₂₈ClFeN₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012152 CAS-Nummer: 16456-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...