Tantal(V)methanolat

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tantal(V)methoxid

18839.06 18839.14
18839.06EA 174 EUR
18839.06 18839.14
Tantal(V)methanolat
Tantal(V)methanolat

CAS No.: 865-35-0 Molecular Formula: C5H15O5Ta Formula Weight: 336.12 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 50° Boiling Point: 189°/10mm UN No.: 3181 MDL No.: MFCD00058817

Formel: C₅H₁₅O₅Ta
Siedepunkt: 189 °C (10 mmHg)
Schmelzpunkt: 50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058817
CAS-Nummer: 865-35-0
UN: 3181
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR