Tantal(V)methanolat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tantal(V)methoxid
18839.06
18839.14
18839.06EA
174
EUR
18839.06
18839.14
Tantal(V)methanolat
Tantal(V)methanolat
CAS No.: 865-35-0
Molecular Formula: C5H15O5Ta
Formula Weight: 336.12
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 50°
Boiling Point: 189°/10mm
UN No.: 3181
MDL No.: MFCD00058817
Formel:
C₅H₁₅O₅Ta Siedepunkt: 189 °C (10 mmHg) Schmelzpunkt: 50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058817 CAS-Nummer: 865-35-0 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...