Supelco® Organic Reference Standard, F.A.M.E. Mix, C20:1-C20:5 Unsaturates
Lieferant: Merck
18913-1AMP
18913-1AMPEA 565 EUR
18913-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix, C20:1-C20:5 Unsaturates
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Verpackung: 50 mg ampule contains approximately equal amounts by weight of: cis-11-Eicosenoic acid aethyl ester (C20:1) cis-1114-Eicosadienoic acid methyl ester (C20:2) cis-111417-Eicosatrienoic acid methyl ester (C20:3) cis-581114-Eicosatetraenoic acid methyl ester (C20:4)
cis-58111417-Eicosapentaenoic acid methyl ester (C20:5)