Supelco® Organic Reference Standard, F.A.M.E. Mix
Lieferant: Merck
18916-1AMP
18916-1AMPEA
90.5
EUR
18916-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix
Standards
Organische Standards
Verpackung: 100 mg ampule contains the following with the weight percentages as indicated. Stearic acid methyl ester (C18:0) 10% Oleic acid methyl ester (C18:1cis-9) 20% Elaidic acid methyl ester (C18:1trans-9) 20% Linoleic acid methyl ester (C18:2cis-912) 20%
Linolelaidic acid methyl ester (C18:2trans-912) 20%
Arachidic acid methyl ester (C20:0) 10%