(2S,3S)-(+)-2,3-Butandiol, Supelco®
Lieferant: Merck
Synonyme:
(S,S)-(+)-2,3-Butylenglykol, (S,S)-(+)-2,3-Butylenglycol, (2S,3S)-(+)-2,3-Butylenglykol, (2S,3S)-(+)-2,3-Dihydroxybutan, L(+)-2,3-Butandiol, (2S,3S)-(+)-2,3-Butylenglycol, (S,S)-(+)-2,3-Butandiol, (S,S)-(+)-2,3-Dihydroxybutan
M-Clarity™ Qualitätsstufe = MQ100
18967-1ML
18967-1MLEA
194
EUR
18967-1ML
(2S,3S)-(+)-2,3-Butandiol, Supelco®
(2S,3S)-(+)-2,3-Butandiol
Formel:
C₄H₁₀O₂ Molecular Weight: 90,12 g/mol Siedepunkt: 179 °C (1013 hPa) Dichte: 0,99 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063648 CAS-Nummer: 19132-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...