2-Sulfobenzoesäurecycloanhydrid ≥90%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 2-sulphobenzoic anhydride, 2-sulphobenzoic acid cyclic anhydride

M-Clarity™ Qualitätsstufe = MQ200

191698-25G
191698-25GEA 151 EUR
191698-25G
2-Sulfobenzoesäurecycloanhydrid ≥90%, Sigma-Aldrich®
2-Sulfobenzoesäurecycloanhydrid
Formel: C₇H₄O₄S
Molecular Weight: 184,17 g/mol
Siedepunkt: 184…186 °C/18 mmHg (Lit.)
Schmelzpunkt: 116 °C
MDL: MFCD00005879
CAS-Nummer: 81-08-3
EINECS: 201-322-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86a7401c63397ed493b250b00cee377b4a91a1b1&vid=86a7401c63397ed493b250b00cee377b4a91a1b1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Grey and Faint, Beige to Beige
Appearance (Form) Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
Titration with NaOH 89.5 - 110.5 %
Infrared Spectrum Conforms to Structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR