4-(Chlormethyl)-2-nitrotoluol ≥97%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 4-(Chloromethyl)-1-methyl-2-nitrobenzene, 4-Methyl-3-nitrobenzyl chloride

192325-25G
192325-25GEA 377 EUR
192325-25G
4-(Chlormethyl)-2-nitrotoluol ≥97%, Sigma-Aldrich®
4-(Chlormethyl)-2-nitrotoluol

4-Methyl-3-nitrobenzyl chloride is used a diagnostic assay manufacturing, hematology, histology.

Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
Siedepunkt: 87…88 °C/0,05 mmHg (Lit.)
Schmelzpunkt: 48,..50 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00044646
CAS-Nummer: 84540-59-0
EINECS: 283-154-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=742fd7b7ba7f92827ce3b2e103c2037fdf6577a1&vid=338a051ec23076bea3612c89e66d394c947405f9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR