Cyclohexanone-2,4-dinitrophenylhydrazone 99, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

192457-1G
192457-1GEA 141 EUR
192457-1G
Cyclohexanone-2,4-dinitrophenylhydrazone 99, Sigma-Aldrich®
Cyclohexanone-2,4-dinitrophenylhydrazone

Useful standard for elemental analysis.

Formel: C₁₂H₁₄N₄O₄
Molecular Weight: 278.27 g/mol
Schmelzpunkt: 159...160 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00001658
CAS-Nummer: 1589-62-4
EINECS: 216-458-1

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Yellow to Orange
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon 51.2 - 52.3%
Nitrogen 19.9 - 20.3%
Purity (TLC) ≥99%
Solubility (Turbidity), 25 mg/ml (2.5%), CHCl₃ Clear
Solubility (Colour) Orange


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR