Bimsstein, BAKER, Laborreagens, J.T.Baker®

Lieferant: Avantor

Synonyme: Bims

1926.1000EA 120 EUR
1926.1000
Bimsstein, BAKER, Laborreagens, J.T.Baker®
Pumice stone
Schmelzpunkt: > 500 °C
MDL: MFCD00148147
CAS-Nummer: 1332-09-8

Bestellen

Spezifikation Testergebnisse

Acid-soluble substances max. 6.0 %
Iron (Fe) passes test
Water Soluble Substances max. 0.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR