Tetrabutylammoniumchlorid Hydrat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
19347.06
19347.14
19347.22
19347.06EA
26.3
EUR
19347.06
19347.14
19347.22
Tetrabutylammoniumchlorid Hydrat
Tetrabutylammoniumchlorid Hydrat
CAS No.: 37451-68-6
Molecular Formula: [CH3(CH2)3]4NCl.xH2O
Formula Weight: 277.92(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 73-75°
MDL No.: MFCD00011635
Formel:
C₁₆H₃₆ClN·xH₂O Schmelzpunkt: 73…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011635 CAS-Nummer: 37451-68-6 EINECS: 214-195-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...