Kupfer(I)bromid-Dimethylsulfid-Komplex
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 54678-23-8
Molecular Formula: CuBr.S(CH3)2
Formula Weight: 205.58
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: Pale brown or pale green
Melting Point: 124-129° dec.
UN No.: 3335
MDL No.: MFCD00043295
Formel:
C₂H₆BrCuS Schmelzpunkt: 124…129 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043295 CAS-Nummer: 54678-23-8 EINECS: 259-292-5 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...