Pyridin ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Azabenzene, Azine
19378.AP
19378.K2
19378.M1
19378.APEA
95.7
EUR
19378.AP
19378.K2
19378.M1
Pyridin ≥99,0% ACS
Pyridine
Purity: 99.0% min
CAS No.: 110-86-1
Molecular Formula: C5H5N
Formula Weight: 79.10
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.978
Refractive Index: 1.5100
Appearance: Colourless to pale yellow
Odour: Unpleasant
Melting Point: -42°
Boiling Point: 115°
Beilstein Reference: 103233
Merck Reference: 14,7970
UN No.: 1282
MDL No.: MFCD00011732
Formel:
C₅H₅N Siedepunkt: 115 °C Schmelzpunkt: –42 °C Dichte: 0,978 Flammpunkt: 20 °C (68 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011732 CAS-Nummer: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 14,7970 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...