6-Hydroxynicotinsäure, Supelco®

Lieferant: Merck

Synonyme: 6-Hydroxypyridin-3-carbonsäure, 6-Hydroxy-3-pyridincarbonsäure

19386-100MG
19386-100MGEA 33.31 EUR
19386-100MG
6-Hydroxynicotinsäure, Supelco®
6-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Schmelzpunkt: 304…306 °C
MDL: MFCD00006277
CAS-Nummer: 5006-66-6
EINECS: 225-682-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=050c70992a4ecf44c0c4c835d1b7ca7b192c8b34&vid=050c70992a4ecf44c0c4c835d1b7ca7b192c8b34&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR