1-Cyclohexene-1-acetic acid
Lieferant: Thermo Fisher Scientific
|
Achtung
|
19462.03
19462.06
19462.14
19462.03EA
38.8
EUR
19462.03
19462.06
19462.14
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
CAS No.: 18294-87-6
Molecular Formula: C8H12O2
Formula Weight: 140.18
Physical Form: Liquid/low melting solid
Density: 1.04
Refractive Index: 1.4775
Melting Point: 33°
Boiling Point: 140-142°/25mm
MDL No.: MFCD00015482
Formel:
C₈H₁₂O₂ Siedepunkt: 140…142 °C (25 mmHg) Schmelzpunkt: 33 °C Dichte: 1,04 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015482 CAS-Nummer: 18294-87-6 EINECS: 242-172-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...