Iron(III) hydroxide, α-phase
Lieferant: Thermo Fisher Scientific
CAS No.: 20344-49-4
Molecular Formula: FeO(OH)
Formula Weight: 88.85
Physical Form: Powder
Density: 3.4-3.9
Odour: Odourless
Solubility: Insoluble in water and alcohol. Soluble in mineral acids
MDL No.: MFCD00064782
Applications: In purifying water, as absorbent in chemical processes, as a pigment, and as a catalyst
Formel:
FeO(OH) Dichte: 3,4…3.9 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064782 CAS-Nummer: 20344-49-4 EINECS: 243-746-4 Merck Index: 14,4025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...