Ammoniumeisen(III)citrat (14,5 - 16,0% Fe), grün, Pulver FCC, J.T.Baker®

Lieferant: Avantor

Synonyme: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

1977-01EA 143.3 EUR
1977-01
Ammoniumeisen(III)citrat (14,5 - 16,0% Fe), grün, Pulver FCC, J.T.Baker®
Ammoniumeisen(III)citrat
Formel: C₆H₅+4y)Fe(x)N(y)O₇
MDL: MFCD00013099
CAS-Nummer: 1185-57-5
EINECS: 214-686-6
Merck Index: 13,00518

Bestellen

Spezifikation Testergebnisse

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical 14.5 - 16.0 %
FCC - Assay (as Fe) 14.5 - 16.0 %
FCC - Identification Passes Test
FCC - Ferric Citrate Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg) ≤1 mg/kg
FCC - Oxalate (C₂O₄) Passes Test
FCC - Sulfate (SO₄) ≤0.3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR