Disulfinblau, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Patentblau VF Natriumsalz, Hydrogen[4-[4-(diethylamino)-2',4'-disulfonatobenzhydryliden]cyclohexa-2,5-dien-1-yliden]diethylammonium, Natriumsalz, Acid Blue1

M-Clarity™ Qualitätsstufe = MQ200

198218-25G
198218-25GEA 105 EUR
198218-25G
Disulfinblau, Sigma-Aldrich®
Disulfinblau

Patent Blue VF has been used in combination with 99mTc phytate to detect sentinel lymph node (SLN) in early stage cervical cancer patients. To detect heat shock protein (HSP) gp96 induced draining lymph nodes (LNs) in mice and to detect the effect of Asn-Gly-Arg (NGR)-tumor necrosis factor (TNF) on tumor perfusion in murine model. It is also used a diagnostic assay manufacturing, hematology, histology.


  • Patent Blue VF dye is a violet colored powder
  • Used in cosmetic coloring

Formel: C₂₇H₃₁N₂NaO₆S₂
Molecular Weight: 566,67 g/mol
Schmelzpunkt: 290 °C
Lagertemperatur: Room Temperature
MDL: MFCD00012119
CAS-Nummer: 129-17-9
EINECS: 204-934-1
Merck Index: 12,09161
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e7244720388bb0817c5a1a2216778b513d83c36&vid=4e7244720388bb0817c5a1a2216778b513d83c36&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d4684321b9d88857feec4f61191f7654f87da7e&vid=13e692a42373b178b1b528f9de5c4b74b89d8a42&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 308 - 314 nm
Extinction coefficient ≥7500
Carbon 28.60 - 75.00 %
Nitrogen 2.40 - 7.40 %
Solubility (Colour) Dark blue to very dark blue and dark blue-green to very dark blue-green

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR