Nigrosine, zertifiziert durch die Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: CI 50420

M-Clarity™ Qualitätsstufe = MQ200

198285-100G 198285-25G
198285-100GEA 105 EUR
198285-100G 198285-25G
Nigrosine, zertifiziert durch die Biological Stain Commission, Sigma-Aldrich®
Nigrosine

Nigrosin,certified by the Biological Stain Commission (Acid black 2; Nigrosin water soluble).


Certified for use in Dorner′s spore stain. Used as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. Used as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi.

Lagertemperatur: Room Temperature
MDL: MFCD00044681
CAS-Nummer: 8005-03-6

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Black
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Wavelength Absorbance 290 - 300 nm
Wavelength 570 - 580 nm
Absorbance c = 0.02 g/L in 50% Ethanol
Biological Stain Commission, Commission Certified Number: Pass
Carbon 40 - 50% Typically
Nitrogen 7 - 9% Typically
TLC, Consistent With Control Conforms
Solubility (Turbidity), c = 0.1% in H2O Clear
Solubility (Colour) Dark Purple


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR