Acridingelb G ≥90% (dye-Basis), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 3,6-Diamino-2,7-dimethylacridine monohydrochloride, CI 46025, Acridine yellow, Acridine Yellow G

M-Clarity™ Qualitätsstufe = MQ100

199508-25G
199508-25GEA 173 EUR
199508-25G
Acridingelb G ≥90% (dye-Basis), Sigma-Aldrich®
Acridingelb G

Acridine Yellow G (AYG) is an organic dye with photophysical properties. It is widely used in optics, medicine and photovoltaic cells.


AYG has been used as a solar photocatalyst to increase biodegradability and remove ferulic acid, a target pollutant from wastewater. It retards the growth of glioblastoma via dual inhibition of epidermal growth factor receptor and protein kinase c kinases.

Formel: C₁₅H₁₅N₃·HCl
Molecular Weight: 273,76 g/mol
MDL: MFCD00012661
CAS-Nummer: 135-49-9
EINECS: 205-194-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff6213c1da9250bccb0da1336f708417df21f87f&vid=40c27241d3edbd5e8ef0d3d3985116c6959f8687&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR