Ethyl orange sodium salt 90% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
4-Diethylaminoazobenzene-4'-sulphonic Acid Sodium Salt
199567-100G
199567-100GEA
336
EUR
199567-100G
Ethyl orange sodium salt 90% (Farbstoffgehalt), Sigma-Aldrich®
Ethyl orange sodium salt
Formel:
C₁₆H₁₈N₃NaO₃S Molecular Weight: 355,39 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007503 CAS-Nummer: 62758-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...